ALX350220M050
ENZOALX350220M050EA
313
EUR
InStock
ENZOALX350220M050
Cafestol ≥98% (par UHPLC)
Cafestol
GST activator.
Inducer of glutathione S-transferases. Has chemoprotective activity, reducing the genotoxicity of several carcinogens.
Soluble in DMSO, 100% ethanol or ethyl acetate.