ALX380088M005
ENZOALX380088M005EA
75.9
EUR
InStock
ENZOALX380088M005
Netropsin dihydrochloride ≥90% (par HPLC)
Netropsin dihydrochloride
Nucleic acid binding ligand.
Nucleic acid binding ligand. Binds to A-T-rich regions of the minor groove of DNA.
Soluble in alcohols or water.