Synonyms:
Sodium 2,6,8-trioxo-1,2,6,7,8,9-hexahydropurin-3-ide, Uric acid sodium, MSU, Uric acid
ALX400047M002
ENZOALX400047M002EA
129
EUR
InStock
ENZOALX400047M002
Monosodium urate, soluble
Monosodium urate
NLRP3 inflammasome activator.
NLRP3/NALP3 inflammasome activator. Specially crystallised and tested for biological activity.
Stimulates the caspase-1 activating NLRP3/NALP3 inflammasome, resulting in the production of active IL-1β and IL-18.
Soluble in PBS or 1M NaOH (25 mg/ml).