Mouse recombinant MegaCD40L™ (soluble) (from CHO cells)
Fournisseur: ENZO LIFE SCIENCES
ALX522120C010
ENZOALX522120C010EA 584.1 EUR InStock
ENZOALX522120C010
Mouse recombinant MegaCD40L™ (soluble) (from CHO cells)
Protéines et Peptides
Highly active
Produced in CHO cells. The extracellular domain of mouse CD40L (CD154) (aa 115-260) is fused at the N-terminus to mouse ACRP30headless (aa 18-111) and a FLAG®-tag.
MegaCD40L™ is a high activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30/adiponectin. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L. It provides a simple and equally potent alternative to [CD40L+enhancer] combinations (Prod. No. ALX-850-075).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité