Synonyms:
(1α,3α,6α,14α,15α,16β)-20-Ethyl-1,6,16-trimethoxy-4-(methoxymethyl)aconitane-3,8,13,14,15-pentol-8-acetate-14-benzoate
ALX550232M025
ENZOALX550232M025EA
113
EUR
InStock
ENZOALX550232M025
Aconitine ≥98% (par HPLC)
Aconitine
Na⁺ channel activator.
A lipid-soluble neurotoxin that acts as a sodium channel modulator.
Binds with high affinity to the open state of the voltage-sensitive sodium channels at site 2, thereby causing a persistent activation of the sodium channels, which become refractory to excitation.
Soluble in 100% ethanol (35 mg/ml); almost insoluble in water (0.3 mg/ml).