ALX550285M050
ENZOALX550285M050EA
82.5
EUR
InStock
ENZOALX550285M050
Pinacidil monohydrate ≥98% (par HPLC)
Pinacidil monohydrate
K⁺ channel activator. Antihypertensive.
Soluble in 100% ethanol (14 mg/ml), DMSO (28 mg/ml) or dilute aqueous acid; insoluble in water.