Anti-CEACAM5 Mouse Monoclonal Antibody
Fournisseur: ENZO LIFE SCIENCES
ALX805077C100
ENZOALX805077C100EA 437 EUR InStock
ENZOALX805077C100
Anti-CEACAM5 Mouse Monoclonal Antibody
Anticorps
CD66e (CEACAM5; CEA) belongs to the carcinoembryonic antigen (CEA) gene family. It encodes a glycosyl phosphatidyl inositol (GPI)-linked glycoprotein with a Mr of 180'000-200'000 which is most strongly expressed on epithelial cells of the fetal and adult colon and to a minor extent on epithelial cells of the stomach and sweat glands, squamous epithelial cells of the tongue, esophagus and cervix. CD66e is used as a tumor marker for early detection of recurrent disease due to its expression in adenocarcinomas of the colon, lung, breast, stomach and pancreas and in mucinous ovarianMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité