Phosphotyrosine detection set
Fournisseur: ENZO LIFE SCIENCES
ALX850025KI01
ENZOALX850025KI01EA 852 EUR InStock
ENZOALX850025KI01
Phosphotyrosine detection set
Systèmes de liaison et de marquage des protéines
This kit offers a set of seven different MAbs specific for tyrosine phosphorylation sites. Phosphorylation patterns in a given cell extract may differ when probed with different antibodies due to sequence specificity. Recognition therefore depends on two criteria: Phosphorylation and surrounding aa motif. If one of the two criteria is not fulfilled, the antibody will not detect the phosphorylation site. Specificity has been determined by epitope mapping using degenerated phosphopeptide libraries.
Informations de commande: Contents: Contains 25µg each of the following MAbs:
Clone 2C8
Clone 3B12
Clone 9H8
Clone 16F4 (Prod. No. ALX-804-174)
Clone 9F1 (Prod. No. ALX-804-669)
Clone 2A5 (Prod. No. ALX-804-670)
Clone 1F9 (Prod. No. ALX-804-671)
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité