Caspase inhibitor set lV
Fournisseur: ENZO LIFE SCIENCES
ALX850239KI01
ENZOALX850239KI01EA 1193.5 EUR InStock
ENZOALX850239KI01
Caspase inhibitor set lV
Suppléments et additifs pour culture cellulaire
Potent, cell permeable synthetic peptides that irreversibly inhibit activities of each of the caspase-family proteases. YVAD, VDVAD, DEVD, LEVD, WEHD, VEID, IETD, LEHD, and AEVD are the recognition sequences for caspase-1, -2, -3, -4, -5, -6, -8, -9, and -10. VAD is a general caspase inhibitor that inhibits all known caspases. Z-FA-FMK is a negative control for FMK inhibitors.
Informations de commande: Contains 25 µl (2 mM) each of the following inhibitors:
- Caspase-1 Inhibitor, Z-YVAD-FMK, yellow cap
- Caspase-2 Inhibitor, Z-VDVAD-FMK, orange cap
- Caspase-3 Inhibitor, Z-DEVD-FMK, red cap
- Caspase-4 Inhibitor, Z-LEVD-FMK, orange cap
- Caspase-5 Inhibitor, Z-WEHD-FMK, clear cap
- Caspase-6 Inhibitor, Z-VEID-FMK, violet cap
- Caspase-8 Inhibitor, Z-IETD-FMK, green cap
- Caspase-9 Inhibitor, Z-LEHD-FMK, blue cap
- Caspase-10 Inhibitor, Z-AEVD-FMK, amber cap
- Caspase-13 Inhibitor, Z-LEED-FMK, yellow cap
- Caspase-Family Inhibitor, Z-VAD-FMK, amber cap
- FMK Negative Control, Z-FA-FMK, clear cap
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité