OXI-TEK TBARS assay kit
Fournisseur: ENZO LIFE SCIENCES
ALX850287KI01
ENZOALX850287KI01EA 931.7 EUR InStock
ENZOALX850287KI01
OXI-TEK TBARS assay kit
Tests Tests cellulaires
The sensitivity of measuring thiobarbituric acid reactive substances (TBARS) has made this assay the method of choice for screening and monitoring lipid peroxidation, which is a major indication of oxidative stress.
Malondialdehyde (MDA) forms a 1:2 adduct with thiobarbituric acid; the adduct can be measured by fluorometry. Biological specimens contain a mixture of TBARS, including lipid hydroperoxides and aldehydes. In practice, TBARS are expressed in equivalent numbers to MDA.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité