Protein carbonyl ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ALX850312KI01
ENZOALX850312KI01EA 997.7 EUR InStock
ENZOALX850312KI01
Protein carbonyl ELISA kit
Tests Tests ELISA
The Protein carbonyl ELISA kit is an antibody-based detection set. It has higher sensitivity, lower background, is less labour-intensive and handles more samples per day as compared to colorimetric assays.
  • Detecting low levels of carbonylated proteins
  • Validated in various sample types
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
  • Assay Time: 4,5 hours
For quantitative determination of carbonylated protein levels in plasma, other body fluids, cell and tissue extracts.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité