Microcystins (adda specific) ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ALX850319KI01
ENZOALX850319KI01EA 1016.4 EUR InStock
ENZOALX850319KI01
Microcystins (adda specific) ELISA kit
Tests Tests ELISA
Polyclonal antibody prepared against the Adda moiety binds to microcystins and nodularins, allowing the congener-independent determination of these toxins and many of its congeners, and does not cross-react with other non-related toxins or compounds. No pre-sample preparation required. Enables simultaneous measurement of multiple samples at reasonable costs.
  • Convenient – no pre-sample preparation required
  • Rapid – analyze 41 duplicate samples in <2,5 hours
  • Assay Time: 1 hour 90 minutes
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité