CV-Cathepsin L detection kit
Fournisseur: ENZO LIFE SCIENCES
BMLAK1270001
ENZOBMLAK1270001EA 434 EUR InStock
ENZOBMLAK1270001
CV-Cathepsin L detection kit
Tests Tests cellulaires
Based on the dye cresyl violet and the cathepsin L PheArg peptide recognition sequence, this kit allows the detection of cathepsin L activity in living cells by fluorescence microscopy.
Once cells are grown, staining is completed in just a few minutes. Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes.
Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis.
Informations de commande: Kit contains CV-(FR)2 reagent, Acridine Orange, Hoechst 33342 stain.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité