MMP inhibitor profiling kit
Fournisseur: ENZO LIFE SCIENCES
BMLAK3080001
ENZOBMLAK3080001EA 1090.1 EUR InStock
ENZOBMLAK3080001
MMP inhibitor profiling kit
Tests Tests enzymatiques
The MMP inhibitor profiling kit, fluorometric RED is a complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes, using a quenched fluorogenic substrate OMNIMMP® RED.
The assays are performed in a convenient 96-well microplate format. The compound NNGH is also included as a prototypic control inhibitor.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité