Matrix Metalloproteinase-12 (MMP-12) colorimetric drug discovery kit
Fournisseur: ENZO LIFE SCIENCES
BMLAK4020001
ENZOBMLAK4020001EA 613.8 EUR InStock
ENZOBMLAK4020001
Matrix Metalloproteinase-12 (MMP-12) colorimetric drug discovery kit
Tests Tests enzymatiques
A QuantiZyme™ Assay System. The MMP-12 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-12 (MMP-12, macrophage elastase, metalloelastase) using a thiopeptide as a chromogenic substrate. The assays are performed in a convenient 96-well plate format. Included are active enzyme, assay buffer, a prototypic control inhibitor (NNGH), and a detailed instruction booklet.
Information de livraison: Kit contains MMP-12 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well Microplate and a detailed instruction booklet.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité