Matrix Metalloproteinase-12 (MMP-12) fluorometric drug discovery kit
Fournisseur: ENZO LIFE SCIENCES
BMLAK4030001
ENZOBMLAK4030001EA 644.6 EUR InStock
ENZOBMLAK4030001
Matrix Metalloproteinase-12 (MMP-12) fluorometric drug discovery kit
Tests Tests enzymatiques
A QuantiZyme™ Assay System. The MMP-12 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-12 (MMP-12, metalloelastase, macrophage elastase) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH.
Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format.
Information de livraison: Kit contains MMP-12 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well Microplate and a detailed instruction booklet.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité