Matrix Metalloproteinase-1 (MMP-1) fluorometric drug discovery kit
Fournisseur: ENZO LIFE SCIENCES
BMLAK4050001
ENZOBMLAK4050001EA 613.8 EUR InStock
ENZOBMLAK4050001
Matrix Metalloproteinase-1 (MMP-1) fluorometric drug discovery kit
Tests Tests enzymatiques
A QuantiZyme™ Assay System. The MMP-1 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-1 (MMP-1, interstitial collagenase) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.
Information de livraison: Kit contains MMP-1 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité