DPPIV Drug discovery kit
Fournisseur: ENZO LIFE SCIENCES
BMLAK4990001
ENZOBMLAK4990001EA 467.5 EUR InStock
ENZOBMLAK4990001
DPPIV Drug discovery kit
Tests Tests enzymatiques
The DPPIV Drug Discovery Kit is a complete assay system designed to screen DPPIV inhibitors, a potential therapeutic target, providing enough material to perform at least 96 assays.
DPPIV (DPP4, CD26) is a member of the class of proteases known as prolyl peptidases, which cleave proteins after proline residues and is thought to play roles in diabetes, cancer, and autoimmune diseases, making it a target for drug discovery.
The kit contains both a chromogenic substrate (H-Gly-Pro-pNA; Km=114 µm) and a fluorogenic substrate (H-Gly-Pro-AMC; Km=50 µm).
Cleavage of the p-nitroaniline (pNA) from the colorimetric substrate increases absorbance at 405 nm. The fluorimetric assay is based on the cleavage of 7-amino-4-methylcoumarin (AMC) moiety from the C-terminus of the peptide substrate, which increases its fluorescence intensity at 460 nm.
A DPPIV inhibitor, P32/98 (KI=130 nm12), is included for use as a control. Other DPP enzymes are available for specificity profiling.
Information de livraison: Kit includes DPPIV enzyme (human, recombinant), pNA substrate, Calibration standard (p-nitroaniline), AMC substrate, Calibration standard (7-Amino-4-methylcoumarin), Inhibitor, Assay Buffer, ½-volume clear microplate
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité