FLUOR DE LYS® HDAC8 fluorometric drug discovery kit
Fournisseur: ENZO LIFE SCIENCES
BMLAK5180001
ENZOBMLAK5180001EA 843 EUR InStock
ENZOBMLAK5180001
FLUOR DE LYS® HDAC8 fluorometric drug discovery kit
Tests Tests cellulaires
A Fluor-de-Lys® fluorescent assay system. The HDAC8 Fluorescent Activity Assay/Drug Discovery Kit is a complete assay system designed to measure the lysyl deacetylase activity of the recombinant human HDAC8 included in the kit. The kit is ideal for chemical library screening for candidate inhibitors or activators or kinetic assay of the enzyme under varying conditions. The Fluor de Lys® HDAC8 assay is based on the Fluor de Lys® Substrate and Fluor de Lys® Developer combination. The assay procedure has two steps. First, the Fluor de Lys® Substrate, which comprises an acetylated lysine side chain, is incubated with HDAC6. Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the Fluor de Lys®Developer produces a fluorophore.
  • Useful for inhibitor screening or characterizing enzyme kinetics
  • Includes optimal substrate selected from a panel of acetylated sites in p53 and histones
  • Supplied with enough recombinant enzyme for 96 assays (1 x 96-well plate)
Information de livraison: Kit contains HDAC8 (human) (recombinant), Fluor de Lys®-HDAC8 substrate and developer II, Assay buffer, Trichostatin A (HDAC inhibitor), White and clear ½-vol. 96-well plates.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité