BMLAK5320001
ENZOBMLAK5320001EA
869
EUR
InStock
ENZOBMLAK5320001
HDAC/SIRT chemiluminescent drug discovery kit, CHEMILUM DE LYS®
Tests
Tests enzymatiques
The HDAC/SIRT chemiluminescent drug discovery kit is a complete assay system designed to measure histone deacetylase (HDAC) and sirtuin activity in cell or nuclear extracts, immunoprecipitates or purified enzymes.
- High specificity assay eliminates false positives or negatives
- Detects without any of the side effects
- Superior signal-to-noise ratio with no interference from cell extract detergents
- Delivers consistent results from a validated system
The HDAC Chemiluminescent activity assay is based on the CHEMILUM DE LYS® substrate and developer combination. The CHEMILUM DE LYS® system (chemiluminescent histone deAcetylase lysyl substrate/developer) is a highly sensitive and convenient alternative to radiolabeled, acetylated histones or peptide/HPLC methods for the assay of histone deacetylases.
The assay procedure has three steps. Firstly, the CHEMILUM DE LYS® substrate, which comprises an acetylated lysine side chain, is incubated with a sample containing HDAC activity (HeLa nuclear or other extract, purified enzyme, bead-bound immunocomplex, etc.). Deacetylation of the substrate sensitises the substrate so that, in the second step, treatment with the CHEMILUM DE LYS® Developer followed by Enhancer produces light. The reaction is luciferase-free.
Information de livraison: Kit contains Nuclear extract from HeLa cells (human cervical cancer cell line), CHEMILUM DE LYS® substrate, CHEMILUM DE LYS® developer concentrate (20X), trichostatin A (HDAC inhibitor), NAD (sirtuin substrate), nicotinamide (sirtuin inhibitor), HDAC assay buffer, developer buffer, CHEMILUM DE LYS® enhancer part A, CHEMILUM DE LYS® enhancer part B and a half volume white microplate.