Calcineurin cellular activity assay kit
Fournisseur: ENZO LIFE SCIENCES
BMLAK8160001
ENZOBMLAK8160001EA 488.4 EUR InStock
ENZOBMLAK8160001
Calcineurin cellular activity assay kit
Tests Tests cellulaires
A complete colorimetric assay kit for measuring cellular calcineurin (PP2B) phosphatase activity. It employs a convenient 96-well plate format with all reagents necessary for measuring calcineurin (PP2B) phosphatase activity in tissue/cellular extracts PLUS human recombinant calcineurin is included as a positive control.
The RII phosphopeptide substrate, supplied with this kit is the most efficient and selective peptide known for calcineurin. The detection of free-phosphate released is based on the classic malachite green assay.
Information de livraison: Kit includes Calcineurin enzyme, Calmodulin (human, recombinant), Substrate (RII phosphopeptide), 2x assay buffer, EGTA Buffer, Lysis Buffer, Protease inhibitor cocktail, Biomol Green™ Reagent, Phosphate standard, Okadaic acid, Desalting column and resin, Half Volume microplate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité