BMLCA1010001
ENZOBMLCA1010001EA
130.9
EUR
InStock
ENZOBMLCA1010001
4-Bromo-A-23187 ≥98% (par CCM) ionophore
4-Bromo-A-23187
Calcium ionophore, non-fluorescent.
A highly selective calcium ionophore which is a useful tool for increasing intracellular Ca²⁺ concentration. A nonfluorescent analog of A-23187 and therefore useful in the presence of fluorescent probes.
Soluble in dichloromethane (clear colorless solution at 10 mg/ml).