BMLCN1050005
ENZOBMLCN1050005EA
519
EUR
InStock
ENZOBMLCN1050005
L-858051 ≥92% (par HPLC)
L-858051
Adenylate cyclase activator.
A water-soluble forskolin derivative, retaining adenylate cyclase stimulatory activity (EC₅₀ = 10 µM).
Soluble in water (>10 mg/ml).