BMLCR1100025
ENZOBMLCR1100025EA
360.8
EUR
InStock
ENZOBMLCR1100025
Pravadoline ≥98% (par CCM)
Pravadoline
Cannabinoid receptor agonist.
Pravadoline is a novel analgesic agent whose mechanism of action includes inhibition of cyclooxygenase and cannabinoid receptor agonist activity.
Soluble in DMSO.