BMLD5150050
ENZOBMLD5150050EA
202.4
EUR
InStock
ENZOBMLD5150050
5(S),15(S)-DIHETE ≥99% (par HPLC)
5(S),15(S)-DIHETE
Arachidonic acid metabolite.
Biosynthesis in human eosinophils. Enhances neutrophil degranulation. Potent chemotaxin for human eosinophils.