BMLEA1270025
ENZOBMLEA1270025EA
476
EUR
InStock
ENZOBMLEA1270025
Phaclofen ≥98% (par HPLC), solide blanc
Phaclofen
GABAB receptor antagonist.
Selective GABAB receptor antagonist. Antagonises the postsynaptic action of baclofen.
Soluble in water, methanol or DMSO.