BMLEI1690010
ENZOBMLEI1690010EA
86.9
EUR
InStock
ENZOBMLEI1690010
(±)4-Hydroxydebrisoquin ≥99%, solide blanc
(±)4-Hydroxydebrisoquin
Major metabolite of debrisoquin.
Useful indicator for cytochrome P-450 polymorphism. For analytical methods.
Soluble in 1 M HCl.