BMLFA0280005
ENZOBMLFA0280005EA
159.5
EUR
InStock
ENZOBMLFA0280005
2-Arachidonoylglycerol ≥98% (par HPLC)
2-Arachidonoylglycerol
Cannabinoid receptor ligand.
Endogenous ligand for the CB₁ receptor (CB₁: Ki=58.3 nM); CB₂: Ki>3 µM) present in high levels in the CNS.
Soluble in DMSO or 100% ethanol.