BMLFA0320050
ENZOBMLFA0320050EA
401.5
EUR
InStock
ENZOBMLFA0320050
N-Arachidonyl-2-chloroethylamide ≥98% (par CCM)
N-Arachidonyl-2-chloroethylamide
Cannabinoid receptor ligand.
Non-pungent, potent and highly selective agonist of the cannabinoid CB₁ receptor (Ki=1.4 nM). Displays >1’400-fold selectivity over CB₂ receptors (Ki=3.1 µM). Active in vivo.
Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml).