Eicosapentaenoyl ethanolamide ≥98% (par CCM)
Fournisseur: ENZO LIFE SCIENCES
BMLFA0470025
ENZOBMLFA0470025EA 238 EUR InStock
ENZOBMLFA0470025
Eicosapentaenoyl ethanolamide ≥98% (par CCM)
Eicosapentaenoyl ethanolamide
Synthetic.
Has been shown to inhibit dietary-restriction-induced lifespan extension in wild-type and TOR pathway mutant nematodes. This data indicates that reduced N-acylethanolamide signaling mediates some of the effect of dietary-restriction-induced lifespan extension and that EPEA, specifically, acts as a metabolic signal that couples nutrient availability with growth and lifespan. EPEA has also been shown to have anti-proliferative effects and anti-inflammatory properties. Compound is subject to oxidation. Protect from air.
Numéro CAS: 109001-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32e71d53e191839537f5d0f911f1dc8b12f5c4b7&vid=545828bb6d8596e90dc3eeac70ba8447648c0af0&prid=web&clid=SAPDAM
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité