BMLFR1010100
ENZOBMLFR1010100EA
161
EUR
InStock
ENZOBMLFR1010100
u-83836e ≥98% (par HPLC), solide blanc à blanc cassé
u-83836e
Antioxidant.
The lazaroid prevents hydrogen peroxide-induced F₂ isoprostane production in renal proximal tubular cells and reduces secondary brain injury in a rabbit model of cryogenic trauma.
Soluble in water (30 mg/ml) or ethanol (>50 mg/ml).