BMLGR1030005
ENZOBMLGR1030005EA
156.2
EUR
InStock
ENZOBMLGR1030005
4-Hydroxyphenylretinamide ≥97% (par CCM)
4-Hydroxyphenylretinamide
Retinoic acid receptor ligand.
An analog of all trans retinoic acid that shows antiproliferative activity on cultured human breast cancer cells. Highly selective activator of retinoid receptors. Induces apoptosis in malignant hemopoietic cell lines.
Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml).