BMLGR1060005
ENZOBMLGR1060005EA
117.7
EUR
InStock
ENZOBMLGR1060005
Methoprene acid ≥98% (par CCM)
Methoprene acid
Retinoid X receptor ligand.
Methoprene acid is an analog of the insect growth regulator, juvenile hormone. It binds directly to RXR and is a transcriptional activator in both insect and mammalian cells. It is RXR specific and does not activate the RAR pathway.
Soluble in DMSO or methanol.