Synonyms:
(2E,4E,6E,8E)-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
BMLGR1080010
ENZOBMLGR1080010EA
164
EUR
InStock
ENZOBMLGR1080010
Acitretin ≥98% (par CCM)
Acitretin
Retinoic acid receptor ligand.
Acitretin is a synthetic aromatic analog of all trans retinoic acid with increased stability in vivo. It is the active free acid form of the antipsoriatic drug etretinate. Displays antiproliferative effect and induces apoptosis in kaposi's sarcoma cells.
Soluble in DMSO.