BMLJ1250005
ENZOBMLJ1250005EA
435
EUR
InStock
ENZOBMLJ1250005
α-Hydroxyalprazolam ≥98% (par CCM)
alpha-Hydroxyalprazolam
Metabolite of alprazolam.
Soluble in DMSO (2.5 mg/ml) or ethanol (2.5 mg/ml).
Attention: Store, as supplied, at room temperature for up to 1 year. Store solutions at -20 °C for up to 3 months.