BMLKC1250500
ENZOBMLKC1250500EA
394
EUR
InStock
ENZOBMLKC1250500
Minoxidil ≥98% (par HPLC)
Minoxidil
K⁺ channel activator.
A selective ATP-sensitive K⁺ (KATP) channel opener. It is metabolised to the more active sulphate in vivo. A clinically useful anti-alopecia. Antihypertensive. Stimulates renin secretion. Potent vascular smooth muscle relaxant.
Soluble in 100% ethanol (29 mg/ml), DMSO (6.5 mg/ml) or water (2.2 mg/ml).