Recombinant Charybdotoxin
Fournisseur: ENZO LIFE SCIENCES
BMLKC1680100
ENZOBMLKC1680100EA 455.4 EUR InStock
ENZOBMLKC1680100
Recombinant Charybdotoxin
Protéines et Peptides
K⁺ channel blocker
Charybdotoxin was originally isolated from the venom of the Israeli scorpion Leiurus quinquestriatus hebraeus. Charybdotoxin blocks KCa1.1 (large conductance Ca²⁺ activated K⁺, Slo) channels in nanomolar concentrations as as well as Kv1.2 (Kd, 14 nM) and Kv1.3 (Kd, 2.6 nM) channels.). However, experiments with cloned KCa1.1 channels demonstrate the strong effect of the slob subunits on the potency of block by Charybdotoxin. This product is a recombinant peptide expressed in and extracted from E. coli.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité