BMLKC1700010
ENZOBMLKC1700010EA
110
EUR
InStock
ENZOBMLKC1700010
N-(4-Phenylsulphonylphenyl)-3,3,3-trifluoro-2-hydroxy-2-methylpropanamide ≥98% (par CCM)
N-(4-Phenylsulphonylphenyl)-3,3,3-trifluoro-2-hydroxy-2-methylpropanamide
K⁺ channel activator.
A unique and selective ATP-sensitive K⁺ channel (KATP) opener (EC50=0.5 μM) which is short acting. Completely abolishes overactivity in a rat model of overactive bladder.
Soluble in DMSO (>25 mg/ml) or ethanol (>25 mg/ml).