BMLKI5900050
ENZOBMLKI5900050EA
244
EUR
InStock
ENZOBMLKI5900050
FLUOR DE LYS®-succinyl, desuccinylase substrate 5 mM ≥95% (par HPLC)
FLUOR DE LYS®-succinyl, desuccinylase substrate
The FLUOR DE LYS®-Succinyl, desuccinylase substrate offers a highly sensitive and convenient alternative to radiolabeled, succinylated histones or peptide/HPLC methods for the assay of desuccinylases such as SIRT5.
Can be used for the study of desuccinylase kinetics, regulation, and inhibitor sensitivity. Ideal for drug discovery and HTS applications.