BMLL1000005
ENZOBMLL1000005EA
64.9
EUR
InStock
ENZOBMLL1000005
1-O-Hexadecyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥98% (par CCM)
1-O-Hexadecyl-2-O-acetyl-sn-glycero-3-phosphocholine
PAF receptor ligand.
Platelet activating factor (PAF) with a C16 alkyl group. Initiates receptor-mediated responses in platelets and a variety of other cells. Rabbit platelet PAF receptor KD=0.1 nM. EC₅₀ for stimulation of phosphoinositide metabolism in hippocampal cell line 1.2-1.5 nM.
Soluble in 100% ethanol (>100 mg/ml). Soluble in water (>100mg/ml) after sonication for 1 minute at 40 °C.