BMLNA1340100
ENZOBMLNA1340100EA
86.9
EUR
InStock
ENZOBMLNA1340100
Lidocaine-N-ethyl bromide ≥98% (par CCM), solide blanc
Lidocaine-N-ethyl bromide
Na+ channel blocker
Quaternary derivative of lidocaine (Prod. No. ALX-550-251). Blocks voltage-gated Na+ channels. Effectively blocks cardiac Na+ channels when applied from either side of the membrane but blocks neuronal Na+channels only from the intracellular side. Externally applied QX-314 was shown to inhibit high frequency activity in sensory nerves
Soluble in water or DMSO.