BMLNA1390001
ENZOBMLNA1390001EA
89.1
EUR
InStock
ENZOBMLNA1390001
Bupivacaine hydrochloride monohydrate ≥98% (par UHPLC), solide blanc
Bupivacaine hydrochloride monohydrate
Sodium channel blocker. Clinically useful local anesthetic.
Soluble in water (5 mg/ml).