BMLNA1410010
ENZOBMLNA1410010EA
315.7
EUR
InStock
ENZOBMLNA1410010
Batrachotoxin ≥90% (par HPLC, TLC)
Batrachotoxin
Na+ channel activator
Batrachotoxin is a potent neurotoxin found in the skin of various poisonous frogs. BTX binds specifically to voltage-gated Na+ channels causing steady state activation, preventing all forms of inactivation, slowing deactivation and altering ion selectivity.
Soluble in DMSO, methanol or 100% ethanol.