BMLNO1090001
ENZOBMLNO1090001EA
161
EUR
InStock
ENZOBMLNO1090001
Salvinorin A ≥98% (par HPLC)
Salvinorin A
Potent naturally occuring diterpenoid κ1-opioid selective agonist. Displays high affinity at κ1-opioid receptors (Ki=4.3 nM).
Soluble in methanol, pyridine, acetonitrile, and chloroform.