R18 peptide
Fournisseur: ENZO LIFE SCIENCES
BMLP2140001
ENZOBMLP2140001EA 226.6 EUR InStock
ENZOBMLP2140001
R18 peptide
Protéines et Peptides
14-3-3 inhibitor
The peptide sequence, PHCVPRDLSWLDLEANMCLP, named R18, was identified as an unphosphorylated peptide that binds to 14-3-3 proteins with high affinity (Kd=80 nM). It has been shown to bind to the same amphipathic groove on the surface of 14-3-3 as phosphorylated peptides. R18 can inhibit the formation of the 14-3-3-Raf-1 complex, reduce Raf-1 dependent transcriptional activity in cells and block the activation of ExoS by 14-3-3. It binds to all the 14-3-3 isoforms with equal affinities. This peptide is useful for competitive inhibition experiments. A biotinylated form is available (BML-P225), allowing detection using streptavidin.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité