BMLPE1350005
ENZOBMLPE1350005EA
253
EUR
InStock
ENZOBMLPE1350005
Phorbol-12,13-dibutyrate ≥99% (par HPLC)
Phorbol-12,13-dibutyrate
Widely used PKC activator (10 to 100 ng/ml).
Less potent than phorbol 12-myristate 13-acettate (PMA; TPA), but also less hydrophobic.
Soluble in DMSO (25 mg/ml) or 100% ethanol (20 mg/ml).