Synonyms:
PtdIns(3,4,5)P3, (1,2-dipalmitoyl)-, 1,2-Dipalmitoyl-sn-glycero-3-phospho-1-D-myo-inositol-3,4,5-trisphosphate·7N
BMLPH1070100
ENZOBMLPH1070100EA
229
EUR
InStock
ENZOBMLPH1070100
L-α-Phosphatidylinositol-3,4,5-P3 ·7Na ≥98% (par CCM)
L-α-Phosphatidylinositol-3,4,5-P3 ·7Na
PKC substrate.
Activates Ca²⁺-insensitive PKC isotypes δ, ε and η. Binds to GRP1 via a PH domain.
Soluble in water (10 mg/ml) or in chloroform/methanol/water (1:1:0.3) (>10 mg/ml).