CA-074 Me
Fournisseur: ENZO LIFE SCIENCES
BMLPI1260001
ENZOBMLPI1260001EA 165 EUR InStock
ENZOBMLPI1260001
CA-074 Me
Protéines et Peptides
Potent and selective irreversible cell permeable cathepsin B inhibitor. Methyl ester (Me) is hydrolyzed by intracellular esterases releasing the active inhibitor. Prevents death of CA1 neurons after ischemia following IV delivery after the ischemic insult. In isolated rat osteoclasts it inhibited bone resorption with a maximal effect at 50 µM. This product is only suitable for in vivo and whole cell experiments. The free acid form must be used for in vitro experiments. Can be used to discriminate between cathepsin B and L/S forms.
  • Cathepsin inhibitor
White solid.
Soluble in DMSO.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fef9e8f22a4fa4230f71e5eef5b472d3675ad05&vid=c372c2bdbafa4b99563f1639ef6948469bf6357d&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité