Calcineurin inhibitory peptide, Enzo Life Sciences
Fournisseur: ENZO LIFE SCIENCES
BMLPR1040500
ENZOBMLPR1040500EA 141.9 EUR InStock
ENZOBMLPR1040500
Calcineurin inhibitory peptide, Enzo Life Sciences
Protéines et Peptides
Sequence is calcineurin (CaN) autoinhibitory domain (rat brain CaN aa 457-482). It inhibits calcineurin with an IC50 of 10 µM using [32P]myosin light chain as substrate. Inhibits Mn²⁺-stimulated calcineurin activity but has no effect on Ni²⁺-stimulated activity. Ki=4,4 µM with radiolabeled [Ala97]-RII (81-99) peptide as the phosphatase substrate.
Sequence is calcineurin (CaN) autoinhibitory domain (rat brain CaN aa 457-482). It inhibits calcineurin with an IC50 of 10 µM using [32P]myosin light chain as substrate. Inhibits Mn2+-stimulated calcineurin activity but has no effect on Ni2+-stimulated activity. Ki=4.4 µM with radiolabeled [Ala97]-RII (81-99) peptide as the phosphatase substrate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité