Ubiquitin-AMC
Fournisseur: ENZO LIFE SCIENCES
BMLSE2110025
ENZOBMLSE2110025EA 273.9 EUR InStock
ENZOBMLSE2110025
Ubiquitin-AMC
Protéines et Peptides
High purity industry standard substrate for deubiquitinylating enzymes (DUBs).
Ubiquitin-AMC is a fluorogenic substrate for a wide range of deubiquitinylating enzymes (DUBs), including ubiquitin C-terminal hydrolases (UCHs) and ubiquitin specific proteases (USPs). It is a particularly useful reagent for the study of deubiquitinylating activity where detection sensitivity or continuous monitoring of activity is essential. Ubiquitin-AMC has been shown to be a sensitive substrate for UCH-L3 (Km = 0,039 μM) (Prod. No. BML-UW9745) and for Isopeptidase-T (Km = 0,17-1,4 μM) (Prod. No. BML-UW9690), and is particularly useful for studying deubiquitinylating activity where detection sensitivity or continuous monitoring of activity is essential.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité